
Potassium hydroxide 0.1 mol/l (0.1 N) in aqueous solution, Titripur® volumetric solution, Supelco®
EM1.09112.1000
:
- Pack type:Plastic bottle
- Size:1 L
- Cat. No.:EM1.09112.1000
- Supplier no.:1.09112.1000
:
:56.11 g/mol
:8,II
:M-Clarity™ quality level = MQ100
:215-181-3
:1814
:GHS07
:Ambient
:1310-58-3
:KOH
:MFCD00003553