Order Entry
Puerto Rico
ContactUsLinkComponent
Extech™ Pressure Transducer 150 psi, Flir®, Extech
Extech™ Pressure Transducer 150 psi, Flir®, Extech
Catalog # 89370-214
Supplier:  Avantor
undefined
Extech™ Pressure Transducer 150 psi, Flir®, Extech
Catalog # 89370-214
Supplier:  Avantor

Specifications

  • Description:
    150 psi pressure transducer
  • Cat. No.:
    89370-214
  • Supplier no.:
    PT150

Specifications

About this item

For air or non-corrosive/non-ionized gas. 1/4" PS, 19 teeth per inch with 3.9 feet/1.2 m cable.

  • Compact size, NPT thread
  • Corrosion-resistant brass alloy
  • Linearized voltage output (0 to 100 mV DC)
  • Half year warranty

The Pressure Transducer for 150 psi measurements with Extech Models 89370-210 pressure meter/datalogger and 407495 heavy duty pressure meter with interchangeable transducers.