ContactUsLinkComponent
Extech™ ExStik™ Replacement Refillable pH Electrode Module For Extech PH100/PH110 Meter, Flir®
Catalog # 89030-378
Supplier:  Avantor
undefined
Extech™ ExStik™ Replacement Refillable pH Electrode Module For Extech PH100/PH110 Meter, Flir®
Catalog # 89030-378
Supplier:  Avantor
Specifications
  • Description:
    ExStik™ replacement refillable pH electrode module for Extech waterproof pH meter
  • Cat. No.:
    89030-378
  • Supplier no.:
    PH115
Specifications
About this item
The electrode is a fresh reference solution that maintains a high accuracy.
  • Easy to clean, rugged flat surface Refillable pH electrode
  • Interchangeable with ExStik® Chlorine or ORP meters
  • Complete with protective connector cap
  • ATC via built-in PT100 Ohm sensor
The electrode is a Refillable pH electrode and is easily serviced. The electrode eliminates concern with shelf life. Provides an economical way to refill your electrode.
Certifications: CE Marked.