Order Entry
Puerto Rico
ContactUsLinkComponent
Extech™ Mini Thermo-Anemometer with Built-in Compass, Flir®
Extech™ Mini Thermo-Anemometer with Built-in Compass, Flir®
Catalog # 75853-970
Supplier:  Avantor
undefined
Extech™ Mini Thermo-Anemometer with Built-in Compass, Flir®
Catalog # 75853-970
Supplier:  Avantor

Specifications

  • Description:
    Mini thermo-anemometer with built-in compass
  • Cat. No.:
    75853-970
  • Supplier no.:
    45168CP

Specifications

About this item

Measure Windspeed in ft/min, MPH, m/s, km/h, Knots and Beaufort Force with 10 second averaging function.

  • Pocket sized meter folds up for storage and extends up to 9" when taking measurements
  • Built-in Compass provides wind direction (Head/Cross/Tail Wind calculation) and 360° readings
  • Delta T displays Air Temperature minus Dew Point
  • Max function recalls highest windspeed reading over time
  • Auto power off 5 minutes after last key is pressed with disable feature
  • Water resistant housing floats and drop-tested to 6 feet
  • Replaceable non-corrosive plastic wind vane (impeller)

The Pocket Thermo-Anemometer which measures Windspeed in ft/min, MPH, m/s, km/h, Knots and Beaufort Force with 10 second averaging function. Additional measurements include Temperature, Relative Humidity, Dew Point, Wet Bulb, and Windchill. The built-in compass provides wind direction/ 360° readings and has an easy-to-use calibration procedure for improved accuracy. Its water resistant housing floats and drop-tested to 6 feet. Featuring a replaceable non-corrosive plastic wind vane (impeller) and a tripod mount for hands free use.

Certifications: CE Marked.
Delivery information: Complete with a CR2032 battery and a lanyard.