Order Entry
Puerto Rico
ContactUsLinkComponent
Pectolyase Y-23 (from Aspergillus niger), MP Biomedicals
Pectolyase Y-23 (from Aspergillus niger), MP Biomedicals
Supplier:  MP Biomedicals
Certificates
undefined
Pectolyase Y-23 (from Aspergillus niger), MP Biomedicals
Supplier:  MP Biomedicals

Specifications

  • CAS No.:
    9032-75-1
  • Source:
    Aspergillus niger
  • Storage Conditions:
    2...8 °C
  • Enzyme Name:
    Pectolyase Y-23

Specifications

Frequently Bought Together