Order Entry
Puerto Rico
ContactUsLinkComponent
Phospholipids C, Wako
Phospholipids C, Wako
Catalog # 11001-736
Supplier:  Wako Life Sciences
CAS Number:  
Phospholipids C, Wako
Catalog # 11001-736
Supplier:  Wako Life Sciences
Supplier Number:  997-01801
CAS Number:  

Specifications

  • Description:
    Phospholipids C
  • Size:
    8 x 50 mL, 2 x 10 mL (kit)
  • Storage Temperature:
    2-10°C
  • Cat. No.:
    11001-736
  • Supplier no.:
    433-36201

Specifications

About this item

The Wako Phospholipids C is an in vitro assay for the quantitative determination of phospholipids in serum.

  • Phospholipid specific enzymatic assay
  • Measures Sphingomyelin, Lecithin, and Lysolecithin
  • Linear up to 1000 mg/dL

Method: Choline oxidase-DAOS

Phospholipids play an important function in the composition of cell membranes and in the emulsification and absorption of fat in the body. Serum phospholipids are formed in the liver, where they become bound to apolipoproteins, which makes them soluble in plasma. The Wako Phospholipids C assay is an enzymatic method utilizing choline oxidase in a reaction which can be measured photometrically. This assay has been shown not to be significantly influenced by coexisting substances such as ascorbic acid and bilirubin.

Composition
Buffer Solution: 8 x 50 mL
Color Reagent: 8 x for 50 mL
Standard Solution: 2 x 10 mL

Caution: For Research Use Only. Not for use in diagnostic procedures.