Order Entry
Puerto Rico
ContactUsLinkComponent
Autokit Glucose, Wako
Autokit Glucose, Wako
Catalog # 11001-718
Supplier:  Wako Life Sciences
CAS Number:  
Autokit Glucose, Wako
Catalog # 11001-718
Supplier:  Wako Life Sciences
Supplier Number:  997-03001
CAS Number:  

Specifications

  • Description:
    Glucose (Autokit)
  • Size:
    2 x 150 mL, 2 x 10 mL (kit)
  • Storage Temperature:
    2-10°C
  • Cat. No.:
    11001-718
  • Supplier no.:
    439-90901

Specifications

About this item

The Wako Autokit Glucose is an in vitro assay for the quantitative determination of glucose in serum, plasma or urine.

Method: Enzymatic

Composition
Buffer Solution: 2 x 150 mL
Color Reagent: 2 x for 150 mL
Standard Solution I: 1 x 10 mL
Standard Solution II: 1 x 10 mL

Caution: For Research Use Only. Not for use in diagnostic procedures.