Order Entry
Puerto Rico
ContactUsLinkComponent
Nylon Bristle and Wood Handle Applicator Brush, Gordon Brush
Nylon Bristle and Wood Handle Applicator Brush, Gordon Brush
Catalog # 22940-756
Supplier:  Gordon Brush
Product available on GSA Advantage®
Small Business Enterprise
Nylon Bristle and Wood Handle Applicator Brush, Gordon Brush
Catalog # 22940-756
Supplier:  Gordon Brush
Supplier Number:  WA12NG-12
Product available on GSA Advantage®
Small Business Enterprise

Specifications

  • Bristle material:
    Nylon
  • Brush head Ø:
    0.012"
  • Handle material:
    Wood
  • Description:
    Upright Brush
  • Brush head width:
    5/16"
  • Cat. No.:
    22940-756
  • No. of rows:
    2×6
  • Handle length:
    415/16"
  • Width:
    1¹⁄₁₆"
  • Supplier no.:
    WA12NG-12
  • Brush head length:
    7/8"
  • Trim length:
    ½"

Specifications

About this item

Small upright applicator brush with 0.012" Nylon bristle and a wooden handle.

  • Works well for getting into tight areas
  • Known as a typewriter brush

Heavy-duty scrubbing and duster brush.