Order Entry
Puerto Rico
ContactUsLinkComponent
 
Nylon Rotary Scrub Brush, Gordon Brush
  22940-664
Product available on GSA Advantage®
Small Business Enterprise
Nylon Rotary Scrub Brush, Gordon Brush
  22940-664
 :  500R
Product available on GSA Advantage®
Small Business Enterprise

 

  • Bristle material:
    Nylon
  • Brush head Ø:
    5"
  • Description:
    Rotary Brush
  • Cat. No.:
    22940-664
  • Supplier no.:
    500R
  • Bristle diameter:
    0.018"

 

 

This Nylon Rotary Scrub Brush can be used with scrubbing machines or flexible shafts.

  • Wood block
  • Nylon bristle

Used extensively for cleaning printed circuit boards.