Tampico Bristle and Curved Wood Handle Plater Brush, Gordon Brush
22940-652
Product available on GSA Advantage®
Small Business Enterprise
- Bristle material:Tampico
- Handle material:Wood
- Description:Plater Brush
- Brush head width:3/4"
- Cat. No.:22940-652
- No. of rows:4×19
- Handle length:127/8"
- Width:1³⁄₁₆"
- Supplier no.:414T
- Brush head length:5 3/8"
- Trim length:1"
This Plater Brush was originally designed for the uniform cleaning and polishing of metal plating.
- Long, slightly curved hardwood handle
- Fill material of natural tampico bristle
An excellent choice for cleaning machines.