ContactUsLinkComponent
Phosphor Bronze Bristle and Plastic Handle Scratch Brush, Gordon Brush
Phosphor Bronze Bristle and Plastic Handle Scratch Brush, Gordon Brush
Catalog # 22940-676
Supplier:  Gordon Brush
Product available on GSA Advantage®
Small Business Enterprise
Phosphor Bronze Bristle and Plastic Handle Scratch Brush, Gordon Brush
Catalog # 22940-676
Supplier:  Gordon Brush
Supplier Number:  21PBM-12
Product available on GSA Advantage®
Small Business Enterprise
Specifications
  • Brush head Ø:
    0.006"
  • Handle material:
    Plastic
  • Description:
    Scratch Brush with Plastic Handle
  • Brush head width:
    1/4"
  • Cat. No.:
    22940-676
  • No. of rows:
    3×7
  • Handle length:
    73/16"
  • Width:
    ⁷⁄₁₆"
  • Supplier no.:
    21PBM-12
  • Brush head length:
    13/8"
  • Trim length:
    ⁷⁄₁₆"
Specifications
About this item
Toothbrush-style scratch brush with a plastic handle designed with a bend for knuckle protection.
  • Ideal for gently cleaning keyboards and narrow areas
Filled with non-sparking phosphor bronze bristle.