Order Entry
Puerto Rico
ContactUsLinkComponent
Stainless Steel Bristle and Acetal Handle Scratch Brush, Gordon Brush
Stainless Steel Bristle and Acetal Handle Scratch Brush, Gordon Brush
Catalog # 22940-580
Supplier:  Gordon Brush
Product available on GSA Advantage®
Small Business Enterprise
Stainless Steel Bristle and Acetal Handle Scratch Brush, Gordon Brush
Catalog # 22940-580
Supplier:  Gordon Brush
Supplier Number:  22SSDG-12
Product available on GSA Advantage®
Small Business Enterprise

Specifications

  • Bristle material:
    Stainless steel
  • Handle material:
    Acetal
  • Description:
    Scratch Brush, Stainless Steel with Plastic (Acetal) Handle
  • Brush head width:
    1/4"
  • Cat. No.:
    22940-580
  • No. of rows:
    2×11
  • Handle length:
    61/4"
  • Supplier no.:
    22SSDG-12
  • Brush head length:
    11/4"
  • Bristle diameter:
    0.003"
  • Trim length:
    ⁷⁄₁₆"

Specifications

About this item

This brush has 0.003" Stainless Steel Bristle with a Delrin (Acetal) handle.

  • Hand-laced with stainless steel wire into its handle for maximum fiber retention
  • Great for removing stubborn dirt, light rust or corrosion on detailed parts
  • Cleans most tarnished metals
  • Can leave a dull finish on soft metals

Autoclavable for cleanroom, sterile, or medical applications.

Certifications: Meets FDA 21 CFR 177.2480 and ASTM D6100 standards.