- Calibration:Up to 3 points
- Temperature Range:0...100 °C (32...212 °F)
- Description:Oakton® PH 550 Benchtop pH Meter Kit
- Temperature Accuracy:±0.5 °C ±1 digit
- Temperature Resolution:0.1/1 °F (0.1 °C)
- Automatic Buffer Recognition:NIST and USA
- Data Logging Capacity:50 groups of data
- Display:LCD
- Temperature Compensation:Automatic or manual from 32...122 °F (0...50 °C)
- mV range:–1999 to 1999 mV
- mV resolution:1
- pH Accuracy:±0.01 pH ±1 digit
- pH Range:0 - 14 pH
- pH Resolution:0.01/0.1
- Cat. No.:76338-508
Economical meter for benchtop pH measurement.
- Automatically recognizes calibration solutions
- Compact design saves valuable benchtop space
- Data logging allows you to store and recall up to 50 groups of data
- IP 54-rated housing is dust- and spill-proof
The PH 550 benchtop pH meter features a microprocessor chip which provides automatic temperature compensation, data storage, parameter setup, and Max/Min reading display. The meter offers a pH range from 0 to 14 and a three-point automatic calibration, with the ability to recognize standard buffer solutions (USA or NIST series). The advanced digital processing technology improves meter response time and measuring accuracy. A stable reading display icon appears when the measuring value is stable.
The 3-in-1 combination pH electrode measures pH and temperature simultaneously. The compact meter includes a detachable electrode holder and three bottles of pH buffer solution. It has a dustproof and spillproof rating of IP54. Connectors are protected by silicone seal caps.