Order Entry
Puerto Rico
ContactUsLinkComponent
Wright-Giemsa stain, Camco® Quik Stain® II
Wright-Giemsa stain, Camco® Quik Stain® II
Catalog # 71130-004
Small Business Enterprise
CAS Number:  
undefined
Wright-Giemsa stain, Camco® Quik Stain® II
Catalog # 71130-004
Supplier Number:  211
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Dispenser bottle
  • Size:
    946 ml (32 fl oz.)
  • Cat. No.:
    71130-004
  • Supplier no.:
    211

Specifications

About this item

 :
MDL Number:MFCD00132593