Manufacturer's Certificate for Julabo Cooling Unit ≥1 kW Cooling Power, Julabo
Catalog # 89422-630
Supplier: JULABO
Specifications
- Description:Manufacturer's Certificate for Julabo Unit with More Than 1 kW Cooling Power at +20 °C
- Cat. No.:89422-630
- Supplier no.:8903035
Specifications
About this item
Manufacturer's certificate measured at +20 °C.
- Production testing certificate
- For systems with >1 kW cooling power