Wright-Giemsa stain, VWR® Quick III™ Set for hematology
Catalog # 10143-224
Supplier: VWR International
CAS Number:
Specifications
- Size:3×90 ml (3×3 oz.)
- Cat. No.:10143-224
Specifications
Specification Test Results
- Quick III Solution I Absorbance:300-340MA @~518nm
- Quick III Solution I pH:6.55-6.65@25°C
- Quick III Solution II Absorbance:440-460MN@~650nm
- Quick III Solution II pH:6.85-6.95@25°C
- Quick III Fixative:Physical Inspection
Specification Test Results
About this item
:
Storage Temperature:Ambient
MDL Number:MFCD00132593