

Hydrochloric acid 6 M HCl (6N) volumetric solution, Fluka™
Catalog # BJX31087-2.5L
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:2.5 L
- Cat. No.:BJX31087-2.5L
Specifications
Specification Test Results
- Appearance (color):complying
- Appearance (form):complying
- APHA:Max. 20
- Assay:5.988 - 6.012 mol/l
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective
Specification Test Results
About this item
:
Density:1.072 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1789
ADR:3,III
:36.46
:GHS07
:GHS05
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324