Order Entry
Puerto Rico
ContactUsLinkComponent
Titration solvent ASTM D-664, LabReady™, Solvent Blend total acid number titration solvent, Burdick & Jackson™
Flammable
Health
Irritant
Titration solvent ASTM D-664, LabReady™, Solvent Blend total acid number titration solvent, Burdick & Jackson™
Catalog # BJWP2148-19P
CAS Number:  
Flammable
Health
Irritant
Titration solvent ASTM D-664, LabReady™, Solvent Blend total acid number titration solvent, Burdick & Jackson™
Catalog # BJWP2148-19P
Supplier Number:  WP2148-19P
CAS Number:  

Specifications

  • Pack type:
    Winpak®
  • Size:
    19 L
  • Cat. No.:
    BJWP2148-19P

Specifications

About this item

 :
Boiling Pt:81,1 °C
Density:0,8215 g/cm3 (25 °C)
Flash Pt:–5 °C
Storage Temperature:Ambient
UN:1993
ADR:3,II
 :GHS02
 :GHS08
 :GHS07