Order Entry
Puerto Rico
ContactUsLinkComponent
Ethyl acetate ≥99.5%, B&J Brand™ for HPLC, for GC (residue analysis), for spectrophotometry, Burdick & Jackson™
Flammable
Irritant
Ethyl acetate ≥99.5%, B&J Brand™ for HPLC, for GC (residue analysis), for spectrophotometry, Burdick & Jackson™
Catalog # BJNS100-20
CAS Number:  
undefined
Flammable
Irritant
Ethyl acetate ≥99.5%, B&J Brand™ for HPLC, for GC (residue analysis), for spectrophotometry, Burdick & Jackson™
Catalog # BJNS100-20
Supplier Number:  NS100-20
CAS Number:  

Specifications

  • Pack type:
    NOWPAK® II, returnable
  • Size:
    20 L
  • Cat. No.:
    BJNS100-20

Specifications

Specification Test Results

  • Electron Capture GC:
    10 ng/L max.
  • Water by Karl Fischer Titration:
    0.03 % max.
  • GC Analysis:
    99.9 % min.
  • Refractive Index (20ºC):
    1.3719 - 1.3727
  • Residue:
    1 mg/L max.
  • UV Absorbance @ 275 nm:
    0.1 AU max.
  • UV Absorbance @ 300 nm:
    0.01 AU max.
  • UV Absorbance @ 325 nm:
    0.005 AU max.
  • UV Absorbance @ 350 nm:
    0.005 AU max.
  • UV Cutoff:
    256 nm max.

Specification Test Results

About this item

 :
 :88.11 g/mol
Boiling Pt:11.5 %(V)
Melting Pt:–84 °C
Density:0.9006 g/cm³ (20 °C)
Flash Pt:–4 °C
UN:1173
ADR:3,II
 :GHS02
 :GHS07
Storage Temperature:Ambient
CAS Number:141-78-6
Formula:CH₃COOC₂H₅
MDL Number:MFCD00009171