Order Entry
Puerto Rico
ContactUsLinkComponent
Acetonitrile with 0,1% (v/v) acetic acid, LabReady™, Solvent Blend for LC-MS, for HPLC, Burdick & Jackson™
Irritant
Health
Flammable
Acetonitrile with 0,1% (v/v) acetic acid, LabReady™, Solvent Blend for LC-MS, for HPLC, Burdick & Jackson™
Catalog # BJLC462-2.5
CAS Number:  
Irritant
Health
Flammable
Acetonitrile with 0,1% (v/v) acetic acid, LabReady™, Solvent Blend for LC-MS, for HPLC, Burdick & Jackson™
Catalog # BJLC462-2.5
Supplier Number:  LC462-2.5
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, GL45
  • Size:
    2.5 L
  • Cat. No.:
    BJLC462-2.5

Specifications

About this item

 :
Boiling Pt:83 °C (main component)
Melting Pt:–46 °C (main component)
Density:0.7770 g/cm³ (25 °C)
Flash Pt:19 °C
Storage Temperature:Ambient
UN:1648
ADR:3,II
 :GHS07
 :GHS08
 :GHS02
CAS Number:75-05-8