


Methanol with 0.1% Ammonium hydroxide, LabReady™, Solvent Blend for LC-MS, for HPLC, Burdick & Jackson™
Catalog # BJLC2149-4
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:4 L
- Cat. No.:BJLC2149-4
Specifications
About this item
:
Boiling Pt:64.4 °C
Density:0.7865 g/cm³ (25 °C)
Flash Pt:11.1 °C
Storage Temperature:Ambient
UN:1230
ADR:3,II
:GHS07
:GHS08
:GHS02
CAS Number:67-56-1