

Methanol : Water (60:40)%, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
Catalog # BJ837-4
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:4 L
- Cat. No.:BJ837-4
Specifications
About this item
:
Boiling Pt:82.78 °C
Density:0.9078 g/cm³ (20 °C)
Flash Pt:18.3 °C
Storage Temperature:Ambient
UN:1230
ADR:3,II
:GHS08
:GHS02
CAS Number:67-56-1
Formula:CH₄O/H₂O
MDL Number:MFCD00004595