2-Propanol:Water (50:50)%, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
BJ703-4
:
- Pack type:Glass bottle
- Size:4 L
- Cat. No.:BJ703-4
:
:82.3 °C (solvent)
:0.9189 g/cm³ (20 °C)
:22.78 °C
:Ambient
:1219
:3,II
:GHS02
:GHS07
:67-63-0