Order Entry
Puerto Rico
ContactUsLinkComponent
Water with 0,1% (v/v) formic acid 0.1% (v/v) in water, CHROMASOLV™, Solvent Blend for LC-MS, Riedel-de Haën™
Water with 0,1% (v/v) formic acid 0.1% (v/v) in water, CHROMASOLV™, Solvent Blend for LC-MS, Riedel-de Haën™
Catalog # BJ576913-4X4L
CAS Number:  
Water with 0,1% (v/v) formic acid 0.1% (v/v) in water, CHROMASOLV™, Solvent Blend for LC-MS, Riedel-de Haën™
Catalog # BJ576913-4X4L
Supplier Number:  576913-4X4L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Size:
    4x 4 L
  • Cat. No.:
    BJ576913-4X4L

Specifications

Specification Test Results

  • Contains:
    0.1 %(v/v) formic acid

Specification Test Results

About this item

 :
Density:0.998 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-791-2
CAS Number:64-18-6
MDL Number:MFCD00011332