


Acetonitrile : Water (80:20)%, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
Catalog # BJNP460-20
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:HDPE NOWPAK®
- Size:20 L
- Cat. No.:BJNP460-20
Specifications
About this item
:
Boiling Pt:66.1 °C
Density:0.8298 g/cm³ (25 °C)
Flash Pt:16 %(V) (main component)
Storage Temperature:Ambient
UN:1648
ADR:3,II
:GHS08
:GHS07
:GHS02
CAS Number:75-05-8
:7732-18-5