


Acetonitrile : Water (50:50)% (w/w), LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
Catalog # BJ457-4
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:4 L
- Cat. No.:BJ457-4
Specifications
About this item
:
Boiling Pt:82 °C (solvent)
Density:0.7857 g/cm³ (20 °C) (pure substance)
Flash Pt:3 °C
Storage Temperature:Ambient
UN:1648
ADR:3,II
:GHS08
:GHS07
:GHS02
CAS Number:75-05-8