

Methanol with 0,1% (v/v) formic acid, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
BJNS445-20
:
- Pack type:NOWPAK® II, returnable
- Size:20 L
- Cat. No.:BJNS445-20
:
:64.7 °C (main component)
:–97.68 °C (main component)
:0.792 g/cm³ (20 °C) (main component)
:11 °C
:1230
:3,II
:GHS02
:GHS08
:Ambient
:64-18-6