

Methanol ≥99.9%, CHROMASOLV™ for capillary GC analysis, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:67-56-1
:
Formula:CH₃OH
MDL Number:MFCD00004595
:32.04 g/mol
Boiling Pt:64…65 °C (1.013 hPa)
Melting Pt:–98 °C
Density:0.79 g/cm³ (20 °C)
Flash Pt:11 °C
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
ADR:3,II
:GHS08
:GHS02
Specification Test Results
- Autoignition Temperature:725 °F
- Identity (IR):complying
- Assay (GC):Min. 99.90 %
- Non-volatile matter:Max. 0.0001 %
- Water (Karl Fischer):Max. 0.10 %
- Dichloromethane (GC):Max. 10 µg/l
- Suitability for residue analysis:Max. 2 ng/l
- Pentachloroethane (GC):Max. 10 µg/l
- Organic impurities as toluene (GC-FID):Max. 1 µg/l
- Vapor Density:1.11 (vs air)
- Vapor Pressure:410 mmHg ( 50 °C)
- Refractive Index:n20/D 1.329 (lit.)