

Chloroform 99-99.4% (by GC) stabilized, meets analytical specification of DAB9, BP, Riedel-de Haën™
Catalog # BJ24216-250KG
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Metal drum
- Size:250 kg
- Cat. No.:BJ24216-250KG
Specifications
Specification Test Results
- Appearance of the substance:complying
- Assay (GC):99 - 99.4 %
- Boiling range:60 - 62 °C
- Density (D 20/20):1.479 - 1.484
- Non-volatile matter:Max. 0.001 %
- Water (Karl Fischer):Max. 0.05 %
- Acid. or alk. react. impurities:complying
- Reaction against H₂SO₄:complying
- Chloride (Cl):Max. 0.0001 %
- Free chlorine:complying
- Foreign chlorine compounds:complying
- Aldehydes:complying
- Ethanol (GC):0.6 - 1.0 Gew.%
- Related substances (GC):complying
Specification Test Results
About this item
:
:119.38 g/mol
Boiling Pt:60…62 °C (1.013 hPa)
Melting Pt:–63 °C
Density:1.48 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-663-8
UN:1888
ADR:6.1,III
:Stabilised with ethanol
:GHS07
:GHS08
CAS Number:67-66-3
Formula:CHCl₃
MDL Number:MFCD00000826