Ground Cord, SCS, Desco Industries
Supplier: Desco
Certificates
About this item
Ground Cords easily snap onto mats and runners.
- 15' (4.6 m) ground cord with 10 mm snap
- Connects all static control mats and runners to building ground
Ground cord has a one megohm resistor for mat connection only.