igScript™ RT-PCR Kit, Intact Genomics
About this item
igScript™ RT-PCR kit requires minimal handling during reaction setup and offer consistent and robust RT-PCR reactions.
- Robust and active for cDNA synthesis at temperatures up to 55 °C
- Enhanced efficiency, specificity, and sensitivity
- Compatible with all real-time PCR instruments
- Superior gene expression results under various cycling conditions
- Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product
- 5X igScript™ master mix
- Oligo d(T)23 VN primer (50 µM)
- Random hexamer primer mix (60 µM)
- Hot start Taq 2X master mix
- Nuclease free water
igScript™ RT-PCR kit combines two powerful mixtures: 5X igScript™ master mix and ii) Hot start Taq 2X master mix with standard buffer for two steps RT-PCR. The two mixtures require minimal handling during reaction setup and offer consistent and robust RT-PCR reactions.
The first strand cDNA synthesis is achieved by using 5X igScript™ master mix which contains igScript™ reverse transcriptase, recombinant RNase inhibitor, dNTPs, an optimized buffer, MgCl₂ and protein stabilizers. IgScript™ reverse transcriptase is a recombinant MMLV reverse transcriptase with reduced RNaseH activity and increased thermostability. The kit also provides two optimized primers and nuclease-free water. An anchored oligo-dT primer forces the primer to anneal to the beginning of the polyA tail and the random hexamer primer mix provides random and consistent priming sites covering the entire RNA templates including both mRNAs and non-polyadenylated RNAs. The kit is highly efficient at producing full-length cDNA from long RNA templates at temperatures between 42 to 55 °C.
The amplification step features a high quality Hot start Taq DNA polymerase which offers higher fidelity and better amplification in a master mix format. RT-PCR product can be generated up to 6 kb.
: All our products are shipped with dry ice via FedEx overnight delivery. US orders received by 1:00 pm (central time, monday-thursday) will typically be shipped on the same day in the US. Orders after 1:00 PM will be shipped on the next business day. Orders received on friday will be ship on the following monday. The shipping and dry ice packaging cost is $40 for your entire order in the 48 contiguous United States.
1 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
igScript™ RT-PCR kit requires minimal handling during reaction setup and offer consistent and robust RT-PCR reactions.
- Robust and active for cDNA synthesis at temperatures up to 55 °C
- Enhanced efficiency, specificity, and sensitivity
- Compatible with all real-time PCR instruments
- Superior gene expression results under various cycling conditions
- Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product
- 5X igScript™ master mix
- Oligo d(T)23 VN primer (50 µM)
- Random hexamer primer mix (60 µM)
- Hot start Taq 2X master mix
- Nuclease free water
igScript™ RT-PCR kit combines two powerful mixtures: 5X igScript™ master mix and ii) Hot start Taq 2X master mix with standard buffer for two steps RT-PCR. The two mixtures require minimal handling during reaction setup and offer consistent and robust RT-PCR reactions.
The first strand cDNA synthesis is achieved by using 5X igScript™ master mix which contains igScript™ reverse transcriptase, recombinant RNase inhibitor, dNTPs, an optimized buffer, MgCl₂ and protein stabilizers. IgScript™ reverse transcriptase is a recombinant MMLV reverse transcriptase with reduced RNaseH activity and increased thermostability. The kit also provides two optimized primers and nuclease-free water. An anchored oligo-dT primer forces the primer to anneal to the beginning of the polyA tail and the random hexamer primer mix provides random and consistent priming sites covering the entire RNA templates including both mRNAs and non-polyadenylated RNAs. The kit is highly efficient at producing full-length cDNA from long RNA templates at temperatures between 42 to 55 °C.
The amplification step features a high quality Hot start Taq DNA polymerase which offers higher fidelity and better amplification in a master mix format. RT-PCR product can be generated up to 6 kb.
: All our products are shipped with dry ice via FedEx overnight delivery. US orders received by 1:00 pm (central time, monday-thursday) will typically be shipped on the same day in the US. Orders after 1:00 PM will be shipped on the next business day. Orders received on friday will be ship on the following monday. The shipping and dry ice packaging cost is $40 for your entire order in the 48 contiguous United States.
Product Family Options
Product Information
- SizeAvailabilityPrice
- 103753-226igScript™ RT-PCR KitigScript™ RT-PCR Kit50 ReactionsDiscontinued-Supplier Number: 4413Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:MarketsourceMarketsource
- 103753-228igScript™ RT-PCR KitSupplier Number: 4415Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:MarketsourceMarketsource