ig-Fusion™ Cloning Kit, Intact Genomics
Propriety ig-Fusion™ cloning technology is a simple, rapid and highly efficient cloning kit which allows to directly clone any PCR products to any linearized expression vector at any site.
- Clone any insert at any site within any vector
- Restriction enzyme and phosphatase free system
- Joining multiple large fragments at once
- Precise insertion at a desired orientation
- Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product
- High efficiency chemically competent cells
PCR fragments can be generated by high fidelity Pfu DNA polymerase or other high fidelity DNA polymerases, with primers having 15 to 18 bases of homology at their linear ends to where the product need to fuse. The linearized vector can be generated by PCR or restriction enzymes. The kit is so robust that multiple DNA fragments can be assembled simultaneously and cloned into one construct in a single reaction step within short times (usually 10 to 30 min) with more than 95% cloning efficiency.
: All our products are shipped with dry ice via FedEx overnight delivery. US orders received by 1:00 pm (central time, monday-thursday) will typically be shipped on the same day in the US. Orders after 1:00 PM will be shipped on the next business day. Orders received on friday will be ship on the following monday. The shipping and dry ice packaging cost is $40 for your entire order in the 48 contiguous United States.