ContactUsLinkComponent
Pfu 2X Master Mix, Intact Genomics
Pfu 2X Master Mix, Intact Genomics
Catalog # 103753-150
Supplier:  Intact Genomics
CAS Number:  
undefined
Pfu 2X Master Mix, Intact Genomics
Catalog # 103753-150
Supplier:  Intact Genomics
Supplier Number:  3326
CAS Number:  
Specifications
  • Description:
    Pfu 2X Master Mix
  • Size:
    200 Reactions (5 ml)
  • Cat. No.:
    103753-150
Specifications
About this item
Pfu 2X master mix is ready to use premix which contains Pfu DNA polymerase, dNTPs, MgCl2 and stabilizers with optimized reaction buffer.
  • Useful for routine PCR, primer extension, colony PCR, genotyping and more
  • Formulated for amplifying long target DNA
  • Supplied as convenient master mix
  • Efficient for amplifying high GC template DNA with magic enhancer
  • Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product
  • Pfu 2X master mix
  • 5X magic enhancer
Pfu DNA polymerase 2X master mix is ready to use premix which contains Pfu DNA polymerase, dNTPs, MgCl₂ and stabilizers with optimized reaction buffer. It has been optimized for routine PCR applications. Pfu DNA polymerase is a heat stable DNA polymerase which has 5' - 3' DNA polymerase and 3' - 5' exonuclease (proof reading) activities. Pfu DNA polymerase retains the high fidelity, sensitivity and processivity with an error rate six-fold lower than Taq DNA polymerase, and significantly lower than the error rates of most other proofreading enzymes or DNA polymerase mixtures.
Delivery: This product is supplied with the unique 5X magic enhancer that enables efficient amplification of GC rich templates up to 84%. All our products are shipped with dry ice via FedEx overnight delivery. US orders received by 1:00 pm (central time, monday-thursday) will typically be shipped on the same day in the US. Orders after 1:00 PM will be shipped on the next business day. Orders received on friday will be ship on the following monday. The shipping and dry ice packaging cost is $40 for your entire order in the 48 contiguous United States.