Order Entry
United States
ContactUsLinkComponent
Hot Start Taq 2X Master Mix, Intact Genomics
Hot Start Taq 2X Master Mix, Intact Genomics
Catalog # 103753-142
CAS Number:  
undefined
Hot Start Taq 2X Master Mix, Intact Genomics
Catalog # 103753-142
Supplier Number:  3296
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Application:
  • Description:
    Hot Start Taq 2X Master Mix
  • Size:
    500 Reactions (12.5 ml)
  • Environmentally Preferable:
  • Cat. No.:
    103753-142

Specifications

About this item

Hot start Taq 2X master mix reduces non-specific amplification during setup stages of the reaction and helps increase PCR specificity and sensitivity.

  • Useful for routine PCR, primer extension, colony PCR and more
  • Supplied as convenient master mix
  • Efficient for amplifying high GC template DNA with magic enhancer
  • Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product
  • 5X magic enhancer

Hot start Taq DNA polymerase 2X master mix is ready to use premix which contains hot start Taq DNA Polymerase, dNTPs, MgCl₂ and stabilizers with optimized reaction buffer. Hot start Taq is a thermostable DNA polymerase that possesses a 5' - 3' polymerase activity and a 5' flap endonuclease activity. Hot start Taq DNA polymerase is chemically modified that leads to complete inactivation of the polymerase until the initial heat activation step at the start of PCR. Hot start PCR reduces non-specific amplification during setup stages of the reaction and helps increase PCR specificity and sensitivity.

Delivery: All our products are shipped with dry ice via FedEx overnight delivery. US orders received by 1:00 pm (central time, monday-thursday) will typically be shipped on the same day in the US. Orders after 1:00 PM will be shipped on the next business day. Orders received on friday will be ship on the following monday. The shipping and dry ice packaging cost is $45 for your entire order in the 48 contiguous United States.