Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
NucSpot®
NucSpot® 470 1000X in DMSO nuclear stain
NucSpot® 470 1000X in DMSO nuclear stain
NucSpot® 470 1000X in DMSO nuclear stain
Supplier:
Biotium
Certificates
Expand image
NucSpot® 470 1000X in DMSO nuclear stain
Supplier:
Biotium
About this item
:
Storage Temperature:
Freezer