Sau DNA Polymerase Large Fragment (from Staphylococcus aureus)
103753-202
:
- Source:Staphylococcus aureus
- Size:1000 units
- Enzyme Name:Sau DNA Polymerase Large Fragment
- Concentration:5 U/µl
- Cat. No.:103753-202
Offers the largest collection of T4 Enzymes and proteins in the industry.
- Extremely high purity
- Variety of uses including RPA optimization
- Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product
- 10X Sau DNA polymerase I reaction buffer
Sau DNA polymerase I, large fragment is a product of the Staphylococcus aureus DNA polymerase I which lacks the N-terminal exonuclease domain (1-293 amino acids). It retains the 5' - 3' polymerase activity of DNA polymerase I but lacks the 5' - 3' exonuclease activity. This large fragment also lacks 3' - 5' exonuclease activity.