Order Entry
Puerto Rico
ContactUsLinkComponent
Bsu DNA Polymerase Large Fragment (from Bacillus subtilis)
Bsu DNA Polymerase Large Fragment (from Bacillus subtilis)
Catalog # 103753-200
Supplier:  Intact Genomics
CAS Number:  
undefined
Bsu DNA Polymerase Large Fragment (from Bacillus subtilis)
Catalog # 103753-200
Supplier:  Intact Genomics
Supplier Number:  3585
CAS Number:  

Specifications

  • Source:
    Bacillus subtilis
  • Size:
    1000 units
  • Enzyme Name:
    Bsu DNA Polymerase Large Fragment
  • Concentration:
    5 U/µl
  • Cat. No.:
    103753-200

Specifications

About this item

Offers the largest collection of T4 Enzymes and proteins in the industry.

  • Extremely high purity
  • Variety of uses including RPA optimization
  • Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product
  • 10X Bsu DNA polymerase I reaction buffer

Bsu DNA polymerase I, large fragment is a product of the Bacillus subtilis DNA polymerase I which lacks the N-terminal exonuclease domain (1-296 amino acids). It retains the 5' - 3' polymerase activity of DNA polymerase I but lacks the 5' - 3' exonuclease activity. This large fragment also lacks 3' - 5' exonuclease activity.