Order Entry
Puerto Rico
ContactUsLinkComponent
T4 gp32 Protein
T4 gp32 Protein
Catalog # 103753-198
CAS Number:  
undefined
T4 gp32 Protein
Catalog # 103753-198
Supplier Number:  3515
CAS Number:  

Specifications

  • Size:
    500 µg
  • Enzyme Name:
    T4 gp32 Protein
  • Concentration:
    5 μg/μl
  • Cat. No.:
    103753-198

Specifications

About this item

Offers the largest collection of T4 Enzymes and proteins in the industry.

  • Extremely high purity
  • Variety of uses including RPA optimization
  • Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product
  • 10X gp32 reaction buffer

T4 gp32 protein is a single-stranded DNA binding protein required for T4 DNA replication, recombination, and repair. It improves the efficiency of reverse transcriptase (RT) during RT-PCR, enhances T4 DNA polymerase activity, as well as increases the yield of PCR product.