Order Entry
Puerto Rico
ContactUsLinkComponent
T4 UvsY Protein (from T4 Phage)
T4 UvsY Protein (from T4 Phage)
Certificates
undefined
T4 UvsY Protein (from T4 Phage)

Specifications

  • Source:
    T4 Phage
  • Enzyme Name:
    T4 UvsY Protein
  • Concentration:
    2 μg/μl

Specifications

Related Information