ContactUsLinkComponent
T4 Polynucleotide Kinase (PNK), Intact Genomics
  103753-188
 :  
undefined
T4 Polynucleotide Kinase (PNK), Intact Genomics
  103753-188
 :  3232
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Size:
    2500 units
  • Enzyme Name:
    T4 Polynucleotide Kinase (PNK)
  • Concentration:
    10 U/µl
  • Cat. No.:
    103753-188
T4 polynucleotide kinase catalyzes the transfer and exchange of Pi from the γ position of ATP to the 5'-hydroxyl terminus of polynucleotides (double-and single-stranded DNA and RNA) and nucleoside 3'-monophosphates.
  • Extremely high purity
  • Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product
  • 10X T4 PNK reaction buffer
T4 polynucleotide kinase catalyzes the transfer of the γ-phosphate from ATP to the 5'-OH group of single and double-stranded DNAs and RNAs, oligonucleotides or nucleoside 3'-monophosphates. In the presence of ADP, T4 PNK exhibits 5'-phosphatase activity and catalyzes the exchange of phosphate group between 5'-P-oligonucleotides/polynucleotides and ATP. The enzyme is also a 3'-phosphatase.
 : All our products are shipped on dry ice.