ContactUsLinkComponent
Taq DNA Ligase (from Thermus aquaticus cloned Escherichia coli), Intact Genomics
  103753-176
 :  
undefined
Taq DNA Ligase (from Thermus aquaticus cloned Escherichia coli), Intact Genomics
  103753-176
 :  3219
 :  
  • Concentration:
    40 U/µl
  • Description:
    Taq DNA Ligase
  • Size:
    10000 Units
  • Cat. No.:
    103753-176
Taq ligase is thermostable DNA ligase from the thermophilic bacterium Thermus aquaticus cloned to E. coli cells.
  • Useful for allele-specific gene detection via LCR and mutagenesis by incorporation of a phosphorylated oligonucleotide
  • Extremely high purity
  • Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product
  • The physical purity of this enzyme is ≥98% as assessed by SDS-PAGE with Coomassie® blue staining
  • 10X Taq DNA ligase buffer with NAD+
Taq DNA ligase catalyzes the formation of a phosphodiester bond in duplex DNA containing adjacent 5'-phosphoryl and 3'-hydroxyl termini, using NAD+ as a cofactor. The ligation will occur only if the oligonucleotides are perfectly paired to the complementary target DNA and have no gaps between them therefore, a single-base substitution can be detected. Taq DNA ligase is active at elevated temperatures (45 to 70°C).
 : All our products are shipped on dry ice.