Hot Start Taq DNA Polymerase
Hot start taq reduces non-specific amplification during setup stages of the reaction and helps increase PCR specificity and sensitivity.
- Useful for routine PCR, primer extension, colony PCR and more
- Efficient for amplifying high GC template DNA with magic enhancer
- Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product.
- Hot start Taq DNA polymerase
- 10X PCR buffer with Mg²⁺
- 5X Magic enhancer
Hot start Taq is a thermostable DNA polymerase that possesses a 5' - 3' polymerase activity and a 5' flap endonuclease activity. Hot start Taq DNA polymerase is chemically modified that leads to complete inactivation of the polymerase until the initial heat activation step at the start of PCR. Hot start PCR reduces non-specific amplification during setup stages of the reaction and helps increase PCR specificity and sensitivity.
: This product is supplied with the unique 10X PCR reaction buffer, containing MgCl₂, which produces a final Mg²⁺ concentration of 1.5 mM, and 5X magic enhancer that enables efficient amplification of GC rich templates up to 84%. All our products are shipped with dry ice. 103753-140 includes 10 mM dNTP.
- Enzyme Name:Hot Start Taq DNA Polymerase
- Concentration:5 U/µl