About this item
Intact Genomics TG1 Phage Display Electrocompetent Cells are suitable for protein expression and preparation of antibody or peptide phage display libraries.
- Intact Genomics (IG) Phage Display Competent Cells Provide the highest transformation efficiency (≥4×10¹⁰ Cfu/µg pUC19 DNA) in the industry
- Amber suppressor strain (supE)
- Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product
Intact Genomics SS320 (MC1061F\’) Phage Display Electrocompetent Cells are suitable for protein expression, general cloning, blue/white screening, M13 phage work and phage display protein expression.
Product Includes SS320 Phage Display electrocompetent cells, pUC19 control DNA and a recovery medium.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Catalog No:
- 103753-128
- 103906-458
- 103753-126
- 103753-132
- Genotypes:
- TG1 phage display electro-competent cells: F’ [traD36 proAB+ lacIq lacZΔM15] supE thi-1 Δ(mcrB-hsdSM)5(rK-mK-) Δ(lac-proAB)
SS320 phage display electro-competent cells: F'[proAB+ lacIq lacZΔM15 Tn10 (tetr)] hsdR mcrA0 araD139 Δ(araA-leu)7697 ΔlacX74 spoT1 galK λe14- galE rpsL thi - TG1 phage display electro-competent cells: F’ [traD36 proAB+ lacIq lacZΔM15] supE thi-1 Δ(mcrB-hsdSM)5(rK-mK-) Δ(lac-proAB)
SS320 phage display electro-competent cells: F'[proAB+ lacIq lacZΔM15 Tn10 (tetr)] hsdR mcrA0 araD139 Δ(araA-leu)7697 ΔlacX74 spoT1 galK λe14- galE rpsL thi - TG1 phage display electro-competent cells: F’ [traD36 proAB+ lacIq lacZΔM15] supE thi-1 Δ(mcrB-hsdSM)5(rK-mK-) Δ(lac-proAB)
SS320 phage display electro-competent cells: F'[proAB+ lacIq lacZΔM15 Tn10 (tetr)] hsdR mcrA0 araD139 Δ(araA-leu)7697 ΔlacX74 spoT1 galK λe14- galE rpsL thi - TG1 phage display electro-competent cells: F’ [traD36 proAB+ lacIq lacZΔM15] supE thi-1 Δ(mcrB-hsdSM)5(rK-mK-) Δ(lac-proAB)
SS320 phage display electro-competent cells: F'[proAB+ lacIq lacZΔM15 Tn10 (tetr)] hsdR mcrA0 araD139 Δ(araA-leu)7697 ΔlacX74 spoT1 galK λe14- galE rpsL thi