About this item
5-alpha electrocompetent E. coli cells are suitable for high efficiency transformation in a wide variety of routine applications such as plasmid isolation, cloning, and subcloning. Mutations in endA1 and recA1 ensure increased plasmid yield and improved plasmid quality.
- 5-alpha electro-competent cells provide the highest transformation efficiency (≥2×1010 Cfu/µg pUC19 DNA) in the industry
- Available as 50 µl vials or 100 μl vials
- Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product
- Includes DH5-alpha electrocompetent cells, pUC19 control DNA and recovery medium
Genomic features a ≥2×1010cfu/µg efficiency with electro-poration a greatly increased plasmid yield and quality due to endA1 mutation a high-efficiency transformation with plasmids 30 kb in size blue/white screening of recombinant clones due to lacZΔM15 and an ensured insert stability due to recA1 mutation.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 103753-120
- 103753-118
- 103753-116
- 103753-114
- Genotypes:
- F – mcrA ∆(mrr-hsdRMS-mcrBC) endA1 recA1 φ80dlacZ∆M15 ∆lacX74 araD139 ∆(ara, leu)7697 galU galK rpsL (StrR) nupG λ
- F – mcrA ∆(mrr-hsdRMS-mcrBC) endA1 recA1 φ80dlacZ∆M15 ∆lacX74 araD139 ∆(ara, leu)7697 galU galK rpsL (StrR) nupG λ
- F – mcrA ∆(mrr-hsdRMS-mcrBC) endA1 recA1 φ80dlacZ∆M15 ∆lacX74 araD139 ∆(ara, leu)7697 galU galK rpsL (StrR) nupG λ
- F – mcrA ∆(mrr-hsdRMS-mcrBC) endA1 recA1 φ80dlacZ∆M15 ∆lacX74 araD139 ∆(ara, leu)7697 galU galK rpsL (StrR) nupG λ