- Volume:6× 100 µl
- Description:igMAX™ DH10B Electro-competent Cells
- Genotypes:F – mcrA ∆(mrr-hsdRMS-mcrBC) endA1 recA1 φ80dlacZ∆M15 ∆lacX74 araD139 ∆(ara, leu)7697 galU galK rpsL (StrR) nupG λ
- Cat. No.:103753-112
igMax™ DH10B derivative electro-competent cells are suitable for demanding cloning situations, such as synthetic bio-applications, BAC cloning, assembling large and multi-DNA fragments, and large construct transformation or cloning difficult targets, requiring the greatest number of transformants possible. Utilizing proprietary manufacturing methods, these cells allow for effective transformation of all large DNA molecules (≥10 kb to 350 kb).
- Extremely large DNA molecules can effectively be transformed at a rate much higher than ANY competitor
- DH10B MAX electro-competent provide the highest transformation efficiency (≥5×1010 cfu/µg pUC19 DNA) in the industry
- Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product
- igMax™ DH10B electro-competent cells, pUC19 control DNA and recovery medium
Genomic features a ≥5×1010 cfu/µg efficiency with electroporation, a 5 to 10×107 for 100 to 150 kb large DNA, high-efficiency transformation with plasmids 30 kb in size blue/white screening of recombinant clones due to lacZΔM15.