Orders LinkContactUsLinkComponent
DH10B ElectroCompetent Cells, Intact Genomics
DH10B ElectroCompetent Cells, Intact Genomics
Marketsource
Supplier:   Intact Genomics
DH10B ElectroCompetent Cells, Intact Genomics
Supplier:  Intact Genomics
Marketsource
Certificates
DH10B ElectroCompetent Cells, Intact Genomics
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
10B (DH10B derivative) electro-competent E. coli cells offer the highest transformation efficiencies of ≥5×1010 cfu/µg plasmid DNA, which are ideal for applications requiring high transformation efficiencies, such as with cDNA or gDNA library construction.
  • DH10B electro-competent cells provide the highest transformation efficiency (≥5×1010 Cfu/µg pUC19 DNA) in the industry
  • Available as 50 µl vials or 100 μl vials
  • Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product
  • Includes DH10B electro-competent cells, pUC19 control DNA and recovery medium
Genomic features aΦ80lacZΔM15 marker provides α-complementation of the β-galactosidase gene with blue/white screening, mcrA genotypic marker and the mcrBC, mrr deletion allows for cloning DNA that contains methylcytosine and methyladenine.
 : All our products are shipped with dry ice.
Specifications for Product Family
Specifications
  • Catalog No:
  • 103753-104
  • 103753-106
  • 103753-108
  • 103753-110
  • Genotypes:
  • F – mcrA ∆(mrr-hsdRMS-mcrBC) endA1 recA1 φ80dlacZ∆M15 ∆lacX74 araD139 ∆(ara, leu)7697 galU galK rpsL (StrR) nupG λ
  • F – mcrA ∆(mrr-hsdRMS-mcrBC) endA1 recA1 φ80dlacZ∆M15 ∆lacX74 araD139 ∆(ara, leu)7697 galU galK rpsL (StrR) nupG λ
  • F – mcrA ∆(mrr-hsdRMS-mcrBC) endA1 recA1 φ80dlacZ∆M15 ∆lacX74 araD139 ∆(ara, leu)7697 galU galK rpsL (StrR) nupG λ
  • F – mcrA ∆(mrr-hsdRMS-mcrBC) endA1 recA1 φ80dlacZ∆M15 ∆lacX74 araD139 ∆(ara, leu)7697 galU galK rpsL (StrR) nupG λ
Specifications
Product Family Options
Product Information
  • Volume
    Availability
    Price
  • 103753-104
    DH10B Electro-competent Cells
    DH10B Electro-competent Cells
    6× 50 µl
    Log in for availability
    Log in for availability
    Each (50µl)
    Each (50µl) - $160.38
    $160.38
    Each (50µl)
       $160.38
    Supplier Number: 1212-12
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    103753-104
    Genotypes
    F – mcrA ∆(mrr-hsdRMS-mcrBC) endA1 recA1 φ80dlacZ∆M15 ∆lacX74 araD139 ∆(ara, leu)7697 galU galK rpsL (StrR) nupG λ
    Marketsource
  • 103753-106
    DH10B Electro-competent Cells
    DH10B Electro-competent Cells
    12× 50 µl
    Log in for availability
    Log in for availability
    Each (50µl)
    Each (50µl) - $303.46
    $303.46
    Each (50µl)
       $303.46
    Supplier Number: 1212-24
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    103753-106
    Genotypes
    F – mcrA ∆(mrr-hsdRMS-mcrBC) endA1 recA1 φ80dlacZ∆M15 ∆lacX74 araD139 ∆(ara, leu)7697 galU galK rpsL (StrR) nupG λ
    Marketsource
  • 103753-108
    DH10B Electro-competent Cells
    DH10B Electro-competent Cells
    6× 100 µl
    Log in for availability
    Log in for availability
    Each (100µl)
    Each (100µl) - $249.23
    $249.23
    Each (100µl)
       $249.23
    Supplier Number: 1214-24
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    103753-108
    Genotypes
    F – mcrA ∆(mrr-hsdRMS-mcrBC) endA1 recA1 φ80dlacZ∆M15 ∆lacX74 araD139 ∆(ara, leu)7697 galU galK rpsL (StrR) nupG λ
    Marketsource
  • 103753-110
    DH10B Electro-competent Cells
    DH10B Electro-competent Cells
    12×100 µl
    Log in for availability
    Log in for availability
    Each (100µl)
    Each (100µl) - $445.38
    $445.38
    Each (100µl)
       $445.38
    Supplier Number: 1214-48
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    103753-110
    Genotypes
    F – mcrA ∆(mrr-hsdRMS-mcrBC) endA1 recA1 φ80dlacZ∆M15 ∆lacX74 araD139 ∆(ara, leu)7697 galU galK rpsL (StrR) nupG λ
    Marketsource