Recovery Media, Intact Genomics
Catalog # 103753-102
Supplier: Intact Genomics
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Volume:6× 12 ml
- Description:Recovery Media
- Cat. No.:103753-102
Specifications
About this item
The recovery medium has superior formulation recommended for all competent cell transformations.
- Achieve the highest possible transformation efficiency from any competent cell
- Enhanced SOC formulation provided with all competent cells
- Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product