Order Entry
Puerto Rico
ContactUsLinkComponent
Orangestick Wood Probes, Menda
Orangestick Wood Probes, Menda
Catalog # 15431-794
Supplier:  Desco
undefined
Orangestick Wood Probes, Menda
Catalog # 15431-794
Supplier:  Desco
Supplier Number:  35616

Specifications

  • Description:
    Orangestick Wood Probe
  • Length:
    17.8 cm (7")
  • Cat. No.:
    15431-794
  • Supplier no.:
    35616

Specifications

About this item

Orangestick wood probes are ideal for use in an ESD protected area.

  • Wood/nylon finish will not mar delicate surfaces
  • Temperature resistant; material will not melt
Packaging: Sold in packages of 100.