Orders LinkContactUsLinkComponent
Distilling Adapters, Process Reactors, Chemglass
Distilling adapter, process reactor, style 'B', 10°, 24/40, 14 mm
  76380-062
 :   Chemglass
 :  CG-1235-V-30
Distilling Adapters, Process Reactors, Chemglass
  76380-062
 :  Chemglass
Distilling Adapters, Process Reactors, Chemglass
Distilling Adapters, Process Reactors, Chemglass
Distilling adapter, process reactor, style 'B', 10°, 24/40, 14 mm
$1,230.00
Each
High quality distillation Adapters are for use with flasks or reaction vessels.
  • Adapter has two top outer joints and a lower inner
  • Adapter has a lower inner joint and two top outer joints
Similar to CG-1235, but with the lower stopcock replaced with a 0 to 14 mm treaded valve having Perfluoro O-rings and a 1/2" O.D. glass distillate takeoff for use with PFA compression tubing fittings. Style “B” with lower angled joints are designed for use with flasks or reaction vessel lids that have a 10° or 20° angled side neck.
  • Description:
    Distilling adapter, process reactor, style 'B', 10°
  • Stopcock size:
    14 mm
  • Joint size:
    24/40