ContactUsLinkComponent
Adapter, Flow Control, Standard Taper Joints, Chemglass
Adapter, Flow Control, Standard Taper Joints, Chemglass
Catalog # 80066-750
Supplier:  Chemglass
Small Business Enterprise
undefined
Adapter, Flow Control, Standard Taper Joints, Chemglass
Catalog # 80066-750
Supplier:  Chemglass
Supplier Number:  CG-1191-02
Small Business Enterprise
Specifications
  • Description:
    Adapter, Flow Control, Chromatography
  • Cat. No.:
    80066-750
  • [SJ] joint no.:
    45/50
  • Supplier no.:
    CG-1191-02
Specifications
About this item
Flow control adapter for use with CG-1188 and CG-1189 chromatography columns along with CG-1190 reservoirs.
  • Adapter has a 0-4 mm Chem-Cap® metering plug